1-Chloro-2-methylpropane ≥98.0%
Supplier: TCI America
C0207-500ML
C0207-25ML
TCC0207-500MLEA
224.96
USD
TCC0207-500ML
TCC0207-25ML
1-Chloro-2-methylpropane ≥98.0%
1-Chloro-2-methylpropane
CAS Number: 513-36-0
MDL Number: MFCD00000954
Molecular Formula: C4H9Cl
Molecular Weight: 92.57
Purity/Analysis Method:
Other Product Names / Synonyms: Isobutyl Chloride
Formula:
C₄H₉Cl Boiling Pt: 70 °C Melting Pt: –131 °C Density: 0.88 (20 °C) Flash Pt: –6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000954 CAS Number: 513-36-0 EINECS: 208-157-9 UN: 1127 ADR: 3,II Merck Index: 13,05155 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...