1-Chlorobutane ≥99.0%

Supplier: TCI America
Danger

C0142-500ML C0142-25ML
TCC0142-500MLEA 51.12 USD
TCC0142-500ML TCC0142-25ML
1-Chlorobutane ≥99.0%
1-Chlorobutane

CAS Number: 109-69-3 MDL Number: MFCD00001009 Molecular Formula: C4H9Cl Molecular Weight: 92.57 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 78 Melting point (°C): -123 Flash Point (°C): -13 Specific Gravity (20/20): 0.89


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement Other Product Names / Synonyms: Butyl Chloride

Formula: CH₃(CH₂)₃Cl
Boiling Pt: 78 °C
Melting Pt: –123 °C
Density: 0.89 (20 °C)
Flash Pt: –13 °C
Storage Temperature: Ambient
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3,II
Merck Index: 13,01559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80b837775fb8cf99752f1a054416fd7aa25c2963&vid=98d417ece07d7567bc2348fa8a773aa8fe706cc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR