3-Chlorobenzoic acid ≥99.0%

Supplier: TCI America
Warning

C0132-25G C0132-500G
TCC0132-025GEA 31 USD
TCC0132-025G TCC0132-500G
3-Chlorobenzoic acid ≥99.0%
3-Chlorobenzoic acid

CAS Number: 535-80-8 MDL Number: MFCD00002491 Molecular Formula: C7H5ClO2 Molecular Weight: 156.57 Purity/Analysis Method: 99.0% (GC,T) Form: Crystal Melting point (°C): 157

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 281
Melting Pt: 122
Density: 1.496 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002491
CAS Number: 535-80-8
EINECS: 208-618-4
Merck Index: 13,02142

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR