1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

B6045-1G
TCB6045-1GEA 126 USD
TCB6045-1G
1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate ≥97.0% (by HPLC, total nitrogen)
1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate
Formula: C₂₁H₂₇BF₄N₂
MDL Number: MFCD02684542
CAS Number: 245679-18-9
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR