SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
Supplier: TCI America
B5834-25ML
TCB5834-25MLEA 46 USD
TCB5834-25ML
SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
Buffers Electrophoresis Buffers Loading Buffers
SDS-PAGE Sample Buffer is specially formulated for protein sample preparation to based on the Laemmli Method of SDS-PAGE system.
This product contains 4% SDS, 20% glycerol, 0.01% bromophenol blue and 0.125 M Tris HCl, pH approx. 6.8. Does not contain 2-Mercaptoethanol.
Caution: Please use after bringing back to normal temperature.