Bismuth(III) iodide, anhydrous ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Bismuth triiodide

B5787-25G B5787-5G
TCB5787-25GEA 107 USD
TCB5787-25G TCB5787-5G
Bismuth(III) iodide, anhydrous ≥98.0% (by titrimetric analysis)
Bismuth (III) iodide
Formula: BiI₃
Boiling Pt: 500 °C
Melting Pt: 408 °C
MDL Number: MFCD00010894
CAS Number: 7787-64-6
EINECS: 232-127-4
UN: 3260
ADR: 8,III
Merck Index: 14,01269

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR