2-Bromo-9,9-di-p-tolyl-9H-fluorene ≥98.0% (by GC)

Supplier: TCI America

B5619-1G B5619-5G
TCB5619-1GEA 156 USD
TCB5619-1G TCB5619-5G
2-Bromo-9,9-di-p-tolyl-9H-fluorene ≥98.0% (by GC)
2-Bromo-9,9-di-p-tolyl-9H-fluorene
Formula: C₂₇H₂₁Br
MDL Number: MFCD16619315
CAS Number: 474918-33-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 182.0 to 186.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR