4-Bromo-9H-fluoren-9-one ≥98.0% (by GC)

Supplier: TCI America
Warning

B5521-1G
TCB5521-1GEA 160 USD
TCB5521-1G
4-Bromo-9H-fluoren-9-one ≥98.0% (by GC)
4-Bromo-9H-fluoren-9-one
Formula: C₁₃H₇BrO
MDL Number: MFCD22988837
CAS Number: 4269-17-4

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR