4,8-Bis(n-octyloxy)benzo[1,2-b:4,5-b']dithiophene-2,6-dicarbaldehyde ≥97.0% (by GC)
Supplier: TCI America
B5484-200MG
TCB5484-200MGEA
179
USD
TCB5484-200MG
4,8-Bis(n-octyloxy)benzo[1,2-b:4,5-b']dithiophene-2,6-dicarbaldehyde ≥97.0% (by GC)
4,8-Bis(n-octyloxy)benzo[1,2-b:4,5-b']dithiophene-2,6-dicarbaldehyde
Formula:
C₂₈H₃₈O₄S₂ |
CAS Number:
1668554-22-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 104.0 to 108.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...