9-Benzoyl-3-bromo-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America

B5471-1G
TCB5471-1GEA 147 USD
TCB5471-1G
9-Benzoyl-3-bromo-9H-carbazole ≥98.0% (by GC)
9-Benzoyl-3-bromo-9H-carbazole
Formula: C₁₉H₁₂BrNO
MDL Number: MFCD30721941
CAS Number: 177775-87-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR