Benzylamine hydrobromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

B5185-1G B5185-5G
TCB5185-1GEA 27 USD
TCB5185-1G TCB5185-5G
Benzylamine hydrobromide ≥98.0% (by HPLC, titration analysis)
Benzylamine hydrobromide
Formula: C₇H₉N·HBr
Melting Pt: 204°C
MDL Number: MFCD00035434
CAS Number: 37488-40-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR