Bis(2,4-dimethylphenyl)amine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5148-1G B5148-200MG
TCB5148-1GEA 310 USD
TCB5148-1G TCB5148-200MG
Bis(2,4-dimethylphenyl)amine ≥98.0% (by GC)
Bis(2,4-dimethylphenyl)amine
Formula: C₁₆H₁₉N
Boiling Pt: 310°C
MDL Number: MFCD16619146
CAS Number: 19616-28-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR