3-Bromo-2-fluorobenzylalkohol ≥98.0% (by GC)

Supplier: TCI America
Warning

B5143-1G B5143-5G
TCB5143-1GEA 27 USD
TCB5143-1G TCB5143-5G
3-Bromo-2-fluorobenzylalkohol ≥98.0% (by GC)
3-Bromo-2-fluorobenzylalkohol
Formula: C₇H₆BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD09842441
CAS Number: 261723-32-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR