Bis(3-biphenylyl)amine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B5048-1G B5048-200MG
TCB5048-1GEA 446 USD
TCB5048-1G TCB5048-200MG
Bis(3-biphenylyl)amine ≥98.0% (by HPLC, total nitrogen)
Bis(3-biphenylyl)amine
Formula: C₂₄H₁₉N
MDL Number: MFCD28129440
CAS Number: 169224-65-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR