4-[Bis(9,9-dimethylfluoren-2-yl)amino]azobenzene ≥98.0% (by HPLC)
Supplier: TCI America
B4796-200MG
TCB4796-200MGEA
328
USD
TCB4796-200MG
4-[Bis(9,9-dimethylfluoren-2-yl)amino]azobenzene ≥98.0% (by HPLC)
4-[Bis(9,9-dimethylfluoren-2-yl)amino]azobenzene
Formula:
C₄₂H₃₅N₃ |
CAS Number:
883554-70-9 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 6.80 to 7.60 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...