4-(tert-Butoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

B4772-1G B4772-200MG B4772-5G
TCB4772-1GEA 91 USD
TCB4772-1G TCB4772-200MG TCB4772-5G
4-(tert-Butoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(tert-Butoxycarbonyl)phenylboronic acid
Formula: C₁₁H₁₅BO₄
MDL Number: MFCD03411946
CAS Number: 850568-54-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR