(3-Bromo-4-methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-Bromo-4-methoxyphenylacetic acid, 2-(3-Bromo-4-methoxyphenyl)acetic acid

B4677-1G B4677-5G
TCB4677-1GEA 56 USD
TCB4677-1G TCB4677-5G
(3-Bromo-4-methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(3-Bromo-4-methoxyphenyl)acetic acid
Formula: C₉H₉BrO₃
MDL Number: MFCD00017538
CAS Number: 774-81-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR