(3-Bromo-4-methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
3-Bromo-4-methoxyphenylacetic acid, 2-(3-Bromo-4-methoxyphenyl)acetic acid
B4677-1G
B4677-5G
TCB4677-1GEA
56
USD
TCB4677-1G
TCB4677-5G
(3-Bromo-4-methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(3-Bromo-4-methoxyphenyl)acetic acid
Formula:
C₉H₉BrO₃ |
MDL Number:
MFCD00017538 CAS Number: 774-81-2 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...