9,9-Bis(4-glycidyloxy-3-methylphenyl)fluorene ≥97.0% (by HPLC)

Supplier: TCI America

B4666-1G B4666-5G
TCB4666-1GEA 131 USD
TCB4666-1G TCB4666-5G
9,9-Bis(4-glycidyloxy-3-methylphenyl)fluorene ≥97.0% (by HPLC)
9,9-Bis(4-glycidyloxy-3-methylphenyl)fluorene
Formula: C₃₃H₃₀O₄
MDL Number: MFCD22494984
CAS Number: 114205-89-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR