Benzo[b]thiophene-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-Benzothiophene-3-carboxylic acid

B4650-1G B4650-200MG
TCB4650-1GEA 155 USD
TCB4650-1G TCB4650-200MG
Benzo[b]thiophene-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Benzo[b]thiophene-3-carboxylic acid
Formula: C₉H₆O₂S
MDL Number: MFCD01846406
CAS Number: 5381-25-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 178.0 to 182.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR