2-Bromo-4,5-difluorophenol ≥98.0%
Supplier: TCI America
Synonyms:
1-Bromo-3,4-difluoro-6-hydroxybenzene
B3845-5G
B3845-1G
TCB3845-5GEA
67
USD
TCB3845-5G
TCB3845-1G
2-Bromo-4,5-difluorophenol ≥98.0%
2-Bromo-4,5-difluorophenol
CAS Number: 166281-37-4
MDL Number: MFCD00070751
Molecular Formula: C6H3BrF2O
Molecular Weight: 208.99
Purity/Analysis Method:
Formula:
C₆H₃BrF₂O Density: 1.84 (20 °C) Flash Pt: 81 °C |
MDL Number:
MFCD00070751 CAS Number: 166281-37-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...