2-Benzylimidazoline ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Tolazoline

B3806-5G
TCB3806-5GEA 71.1 USD
TCB3806-5G
2-Benzylimidazoline ≥97.0% (by GC, titration analysis)
2-Benzylimidazoline

CAS Number: 59-98-3 MDL Number: MFCD00005182 Molecular Formula: C10H12N2 Molecular Weight: 160.22 Purity/Analysis Method: 97.0% (GC,T) Form: Crystal Boiling point (°C): 188 Melting point (°C): 66 Storage Temperature: 0-10°C


Product Categories: Bioscience, Other Pharmaceutical Ingredients Other Product Names / Synonyms: Tolazoline

Formula: C₁₀H₁₂N₂
MW: 160.22 g/mol
Boiling Pt: 188 °C
Melting Pt: 66 °C
MDL Number: MFCD00005182
CAS Number: 59-98-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a3cc628935a2550f98f91dfa4e4ad0e4f9c7664&vid=1a3cc628935a2550f98f91dfa4e4ad0e4f9c7664&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR