5-Bromo-1H-indazole ≥97.0% (by GC)

Supplier: TCI America

B3785-1G B3785-5G
TCB3785-1GEA 56 USD
TCB3785-1G TCB3785-5G
5-Bromo-1H-indazole ≥97.0% (by GC)
5-Bromo-1H-indazole

CAS Number: 53857-57-1 MDL Number: MFCD00839493 Molecular Formula: C7H5BrN2 Molecular Weight: 197.04 Purity/Analysis Method: 97.0% (GC) Form: Crystal Melting point (°C): 132

Formula: C₇H₅BrN₂
Melting Pt: 132 °C
MDL Number: MFCD00839493
CAS Number: 53857-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5be616c9303a4e741fd5fef58bb604c58c536ab&vid=e5be616c9303a4e741fd5fef58bb604c58c536ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR