5-Bromo-1H-indazole ≥97.0% (by GC)
Supplier: TCI America
B3785-1G
B3785-5G
TCB3785-1GEA
56
USD
TCB3785-1G
TCB3785-5G
5-Bromo-1H-indazole ≥97.0% (by GC)
5-Bromo-1H-indazole
CAS Number: 53857-57-1
MDL Number: MFCD00839493
Molecular Formula: C7H5BrN2
Molecular Weight: 197.04
Purity/Analysis Method:
Formula:
C₇H₅BrN₂ Melting Pt: 132 °C |
MDL Number:
MFCD00839493 CAS Number: 53857-57-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...