Benzyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis) for biochemical research

Supplier: TCI America

B3768-25G B3768-5G
TCB3768-25GEA 31.34 USD
TCB3768-25G TCB3768-5G
Benzyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis) for biochemical research
Benzyl 4-hydroxybenzoate

[for Biochemical Research]
CAS Number: 94-18-8
MDL Number: MFCD00016471
Molecular Formula: C14H12O3
Molecular Weight: 228.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 113


Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Benzyl Ester, Benzylparaben

Formula: C₁₄H₁₂O₃
Melting Pt: 113 °C
MDL Number: MFCD00016471
CAS Number: 94-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=465e78d15b20f9c61044becfe90a70cdde4059a4&vid=50685b16fdb40edd96628b1c8b67aecff913422f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR