N-Fmoc-D-glutamic acid 5-tert-butyl ester ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamate, 5-tert-Butyl N-Fmoc-D-glutamate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid 5-tert-Butyl Ester, Fmoc-D-Glu(OtBu)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid
CAS Number: 104091-08-9
MDL Number: MFCD00077055
Molecular Formula: C24H27NO6
Molecular Weight: 425.48
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 92
Specific rotation [a]20/D: 9 deg (C=1, MeOH)
Storage Temperature: 0-10°C
Formula:
C₂₄H₂₇NO₆ MW: 425.48 g/mol Melting Pt: 92 °C |
MDL Number:
MFCD00077055 CAS Number: 104091-08-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...