2-Bromo-6-fluoropyridine ≥98.0% (by GC)
Supplier: TCI America
B3667-1G
B3667-5G
TCB3667-1GEA
38
USD
TCB3667-1G
TCB3667-5G
2-Bromo-6-fluoropyridine ≥98.0% (by GC)
2-Bromo-6-fluoropyridine
CAS Number: 144100-07-2
MDL Number: MFCD03095258
Molecular Formula: C5H3BrFN
Molecular Weight: 175.99
Purity/Analysis Method:
Formula:
C₅H₃BrFN Boiling Pt: 164 °C Melting Pt: 32 °C |
MDL Number:
MFCD03095258 CAS Number: 144100-07-2 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...