4-Bromo-m-cresol ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 14472-14-1
MDL Number: MFCD00079723
Molecular Formula: C7H7BrO
Molecular Weight: 187.04
Purity/Analysis Method:
Other Product Names / Synonyms: 4-Bromo-m-cresol
Formula:
C₇H₇BrO Boiling Pt: 145 °C Melting Pt: 68 °C |
MDL Number:
MFCD00079723 CAS Number: 14472-14-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...