1-Propanol ≥75.0% (by HPLC)
Supplier: TCI America
Synonyms:
Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
B3514-100MG
TCB3514-100MGEA
152.7
USD
TCB3514-100MG
1-Propanol ≥75.0% (by HPLC)
1-Propanol
CAS Number: 207347-46-4
MDL Number: MFCD11042475
Molecular Formula: C58H86N8O8RuS2
Molecular Weight: 1188.56
Purity/Analysis Method:
Product Categories: Materials Chemistry, Solar Cell Research Reagents
Other Product Names / Synonyms: N719 Dye
Formula:
H₃CCH₂CH₂OH MW: 60.1 g/mol Melting Pt: 260 °C |
MDL Number:
MFCD00002941 CAS Number: 71-23-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...