Boc-Aib-OH ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 30992-29-1
MDL Number: MFCD00042973
Molecular Formula: C9H17NO4
Molecular Weight: 203.24
Purity/Analysis Method:
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 2-(Boc-amino)isobutyric Acid, N-Boc-alpha-methylalanine, Boc-Aib-OH
Formula:
C₉H₁₇NO₄ Melting Pt: 120 °C |
MDL Number:
MFCD00042973 CAS Number: 30992-29-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...