2-Bromo-3-nitrobenzoic acid ≥98.0%
Supplier: TCI America
|
Warning
|
B3322-5G
B3322-25G
TCB3322-5GEA
162
USD
TCB3322-5G
TCB3322-25G
2-Bromo-3-nitrobenzoic acid ≥98.0%
2-Bromo-3-nitrobenzoic acid
CAS Number: 573-54-6
MDL Number: MFCD00074899
Molecular Formula: C7H4BrNO4
Molecular Weight: 246.02
Purity/Analysis Method:
Formula:
C₇H₄BrNO₄ MW: 246.02 g/mol Melting Pt: 188 °C |
MDL Number:
MFCD00074899 CAS Number: 573-54-6 EINECS: 209-356-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...