1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate ≥97.0%
Supplier: TCI America
Synonyms:
1,2-Bis(phenylsulphinyl)ethanepalladium(II) acetate, 1,2-Bis(phenylsulfinyl)ethanepalladium(II) acetate, 1,2-Bis(phenylsulfinyl)ethane palladium(II) acetate, 1,2-Bis(phenylsulphinyl)ethane palladium(II) acetate, White Catalyst, Christina White Catalyst
CAS Number: 858971-43-4
MDL Number: MFCD09842752
Molecular Formula: C18H20O6PdS2
Molecular Weight: 502.89
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, C-H Bond Activation Reaction
Other Product Names / Synonyms: White Catalyst
Formula:
C₁₆H₁₈O₄PdS₂ |
MDL Number:
MFCD09842752 CAS Number: 858971-43-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...