3-Bromo-5-fluoropyridine ≥98.0%

Supplier: TCI America

B3244-5G B3244-1G
TCB3244-5GEA 80 USD
TCB3244-5G TCB3244-1G
3-Bromo-5-fluoropyridine ≥98.0%
3-Bromo-5-fluoropyridine

CAS Number: 407-20-5 MDL Number: MFCD04112555 Molecular Formula: C5H3BrFN Molecular Weight: 175.99 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 78 Flash Point (°C): 64 Freezing point (°C): 25

Formula: C₅H₃BrFN
Boiling Pt: 78 °C
Flash Pt: 64 °C
MDL Number: MFCD04112555
CAS Number: 407-20-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=386c6918fa0438e61772a47e71899c1a0b132fab&vid=386c6918fa0438e61772a47e71899c1a0b132fab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR