1-Butyl-3-methylimidazolium methanesulfonate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Danger
|
Synonyms:
1-n-Butyl-3-methylimidazolium methanesulfonate, 1-n-Butyl-3-methylimidazolium methanesulphonate
B3231-25G
B3231-5G
TCB3231-25GEA
120
USD
TCB3231-25G
TCB3231-5G
1-Butyl-3-methylimidazolium methanesulfonate ≥98.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazolium methanesulphonate
Formula:
C₉H₁₈N₂O₃S |
MDL Number:
MFCD06798173 CAS Number: 342789-81-5 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...