1-Butyl-3-methylimidazolium methanesulfonate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

Synonyms: 1-n-Butyl-3-methylimidazolium methanesulfonate, 1-n-Butyl-3-methylimidazolium methanesulphonate

B3231-25G B3231-5G
TCB3231-25GEA 120 USD
TCB3231-25G TCB3231-5G
1-Butyl-3-methylimidazolium methanesulfonate ≥98.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazolium methanesulphonate
Formula: C₉H₁₈N₂O₃S
MDL Number: MFCD06798173
CAS Number: 342789-81-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR