Coomassie brilliant blue G-250
Supplier: TCI America
Synonyms:
Brilliant blue G-250, Serva Blue G, Coomassie Blue G 250, Xylene Brilliant Cyanine G, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulfonatobenzyl)ammonium monosodium salt, Brillant blue G, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt, C.I. 42655, Acid Blue 90, Sodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulphonate, Brilliant indocyanine G, Brilliant Blue G, Sodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulfonate
CAS Number: 6104-58-1
MDL Number: MFCD00078482
Molecular Formula: C47H48N3NaO7S2
Molecular Weight: 854.02
Form: Crystal
Color: Red
Other Product Names / Synonyms: Acid Blue 90, Brilliant Blue G, CBB G-250
Formula:
C₄₇H₄₈N₃NaO₇S₂ MW: 854.04 g/mol |
MDL Number:
MFCD00078482 CAS Number: 6104-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...