β-methasone-21-acetate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
9ɑ-Fluoro-11β,17ɑ,21-trihydroxy-16β-methyl-1,4-pregnadiene-3,20-dione 21-Acetate
CAS Number: 987-24-6
MDL Number: MFCD00867462
Molecular Formula: C24H31FO6
Molecular Weight: 434.50
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Immune System, Inflammation, Allergy Research Reagents, Pregnanes (Glucocorticoids), Steroids
Other Product Names / Synonyms: 9alpha-Fluoro-11beta,17alpha,21-trihydroxy-16beta-methyl-1,4-pregnadiene-3,20-dione 21-Acetate
Formula:
C₂₄H₃₁FO₆ MW: 434.50 g/mol Melting Pt: 215 °C |
MDL Number:
MFCD00867462 CAS Number: 987-24-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...