5-Bromoquinoline ≥98.0%
Supplier: TCI America
B3078-1G
TCB3078-1GEA
51
USD
TCB3078-1G
5-Bromoquinoline ≥98.0%
5-Bromoquinoline
CAS Number: 4964-71-0
MDL Number: MFCD00234481
Molecular Formula: C9H6BrN
Molecular Weight: 208.06
Purity/Analysis Method:
Formula:
C₉H₆BrN Boiling Pt: 280 °C Melting Pt: 46 °C |
MDL Number:
MFCD00234481 CAS Number: 4964-71-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...