N-Benzylglycine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Bzl-Gly-OH·HCl, Bzl-Gly-OH.HCl
B3075-25G
B3075-5G
TCB3075-25GEA
115.46
USD
TCB3075-25G
TCB3075-5G
N-Benzylglycine hydrochloride ≥98.0%
N-Benzylglycine hydrochloride
CAS Number: 7689-50-1
MDL Number: MFCD00156934
Molecular Formula: C9H11NO2
Molecular Weight: 201.65
Purity/Analysis Method:
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Bzl-Gly-OH.HCl
Formula:
C₉H₁₁NO₂·HCl MW: 201.7 g/mol Melting Pt: 228 °C |
MDL Number:
MFCD00156934 CAS Number: 7689-50-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...