3-Bromo-p-toluic acid ≥98.0%
Supplier: TCI America
B3049-25G
B3049-5G
TCB3049-25GEA
78
USD
TCB3049-25G
TCB3049-5G
3-Bromo-p-toluic acid ≥98.0%
3-Bromo-p-toluic acid
CAS Number: 7697-26-9
MDL Number: MFCD00002488
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 3-Bromo-p-toluic Acid
Formula:
C₈H₇BrO₂ Melting Pt: 210 °C |
MDL Number:
MFCD00002488 CAS Number: 7697-26-9 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...