5-Benzylmercaptotetrazole (BMT, BTT) ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B3020-25G
TCB3020-25GEA
200.78
USD
TCB3020-25G
5-Benzylmercaptotetrazole (BMT, BTT) ≥98.0% (by HPLC, titration analysis)
5-(Benzylthio)-1H-tetrazole
CAS Number: 21871-47-6
MDL Number: MFCD00068731
Molecular Formula: C8H8N4S
Molecular Weight: 192.24
Purity/Analysis Method:
Formula:
C₈H₈N₄S Melting Pt: 133 °C |
MDL Number:
MFCD00068731 CAS Number: 21871-47-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...