Benzo[ghi]perylene ≥97.0% (by GC)
Supplier: TCI America
B2983-100MG
TCB2983-100MGEA
278
USD
TCB2983-100MG
Benzo[ghi]perylene ≥97.0% (by GC)
Benzo[ghi]perylene
CAS Number: 191-24-2
MDL Number: MFCD00004135
Molecular Formula: C22H12
Molecular Weight: 276.34
Purity/Analysis Method:
Formula:
C₂₂H₁₂ Boiling Pt: 542 °C Melting Pt: 273 °C |
MDL Number:
MFCD00004135 CAS Number: 191-24-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...