3-Bromo-4-fluoroaniline ≥97.0%

Supplier: TCI America

B2957-5G
TCB2957-5GEA 55 USD
TCB2957-5G
3-Bromo-4-fluoroaniline ≥97.0%
3-Bromo-4-fluoroaniline

CAS Number: 656-64-4 MDL Number: MFCD03788559 Molecular Formula: C6H5BrFN Molecular Weight: 190.02 Purity/Analysis Method: 97.0% (GC,T) Form: Crystal Boiling point (°C): 124 Freezing point (°C): 29

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 124 °C
MDL Number: MFCD03788559
CAS Number: 656-64-4
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73225a53d05ab7219f2f26cbbe9dec1ff67df0b1&vid=73225a53d05ab7219f2f26cbbe9dec1ff67df0b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR