9-Bromoanthracene ≥99.0%

Supplier: TCI America

B2871-25G B2871-100G B2871-5G
TCB2871-25GEA 98 USD
TCB2871-25G TCB2871-100G TCB2871-5G
9-Bromoanthracene ≥99.0%
9-Bromoanthracene

CAS Number: 1564-64-3 MDL Number: MFCD00001243 Molecular Formula: C14H9Br Molecular Weight: 257.13 Purity/Analysis Method: 99.0% (GC,HPLC) Form: Crystal Melting point (°C): 105

Formula: C₁₄H₉Br
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00001243
CAS Number: 1564-64-3
EINECS: 216-359-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f99756e5a5ca473172198600985a7e05e46b2162&vid=08b7566b9c14dfa4810528ef1ea0dbf08bc6b209&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR