5-Bromo-2-hydroxypyrimidine ≥95.0%

Supplier: TCI America

B2848-25G B2848-5G
TCB2848-25GEA 113 USD
TCB2848-25G TCB2848-5G
5-Bromo-2-hydroxypyrimidine ≥95.0%
5-Bromo-2-hydroxypyrimidine

CAS Number: 38353-06-9 Molecular Formula: C4H3BrN2O Molecular Weight: 174.99 Purity/Analysis Method: 95.0% (GC) Form: Crystal Melting point (°C): 235


Other Product Names / Synonyms: 5-Bromo-2-pyrimidinol, 5-Bromo-2(1H)-pyrimidinone

Formula: C₄H₃BrN₂O
Melting Pt: 235 °C
MDL Number: MFCD01318956
CAS Number: 38353-06-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ca430c5e1419c57c61510d0acc9886e43f69240&vid=4ca430c5e1419c57c61510d0acc9886e43f69240&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR