4-Bromoindole-3-carboxyaldehyde ≥96.0% (by GC, total nitrogen)
Supplier: TCI America
B2773-1G
B2773-5G
TCB2773-1GEA
51.3
USD
TCB2773-1G
TCB2773-5G
4-Bromoindole-3-carboxyaldehyde ≥96.0% (by GC, total nitrogen)
4-Bromoindole-3-carboxyaldehyde
CAS Number: 98600-34-1
Molecular Formula: C9H6BrNO
Molecular Weight: 224.06
Purity/Analysis Method:
Other Product Names / Synonyms: 4-Bromo-3-formylindole
Formula:
C₉H₆BrNO Melting Pt: 179 °C |
MDL Number:
MFCD05864695 CAS Number: 98600-34-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...