(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥98.0%

Supplier: TCI America

Synonyms: XANTPHOS, 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene

B2709-1G B2709-5G B2709-25G
TCB2709-1GEA 57.08 USD
TCB2709-1G TCB2709-5G TCB2709-25G
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥98.0%
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)

CAS Number: 161265-03-8 Molecular Formula: C39H32OP2 Molecular Weight: 578.63 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Melting point (°C): 229


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Ligands, Ligands ([2+2+2]Cycloaddition Reaction), Materials Chemistry, Precise Polymerization Reagents Other Product Names / Synonyms: Xantphos

Formula: C₃₉H₃₂OP₂
MW: 578.62 g/mol
Melting Pt: 229 °C
MDL Number: MFCD00233866
CAS Number: 161265-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11b50ece15678fa12902d6db88329dde61957e11&vid=11b50ece15678fa12902d6db88329dde61957e11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR