Bathocuproin ≥99.0% (by HPLC, titration analysis), purified by sublimation

Supplier: TCI America

Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline

B2694-1G B2694-5G
TCB2694-1GEA 354.31 USD
TCB2694-1G TCB2694-5G
Bathocuproin ≥99.0% (by HPLC, titration analysis), purified by sublimation
Bathocuproin

CAS Number: 4733-39-5
MDL Number: MFCD00004972
Molecular Formula: C26H20N2
Molecular Weight: 360.46
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 283


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Highly Purified Reagents, Analytical Science, Phenanthrolines (Chelating Agents)
Other Product Names / Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline

Formula: C₂₆H₂₀N₂
Melting Pt: 283 °C
MDL Number: MFCD00004972
CAS Number: 4733-39-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17474c845dab44cce4189a350cd9237533d79406&vid=9a24e98d5e8fc38cb93a10574e300b2b790a3cb3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR