Cobalt(II) acetylacetonate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Cobalt(II) bis(acetylacetonate), Cobalt (II) bis(2,4-pentanedionato-O,O'), Cobalt (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')cobalt (II)

B2681-25G
TCB2681-25GEA 29.3 USD
TCB2681-25G
Cobalt(II) acetylacetonate ≥97.0% (by titrimetric analysis)
Cobalt (II) acetylacetonate

CAS Number: 14024-48-7
Molecular Formula: C10H14CoO4
Molecular Weight: 257.15
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 173


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Oxidation, Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Acetylacetone Cobalt(II) Salt, Cobalt(II) Acetylacetonate

Formula: C₁₀H₁₄CoO₄
Melting Pt: 173 °C
MDL Number: MFCD00000014
CAS Number: 14024-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e9de8c077f165d048e73ab7eae1cf5b60475713&vid=2f333b8ac959b41514f3eb59e7a114415537beef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR