1,2-Benzisothiazol-3(2H)-one ≥98.0%
Supplier: TCI America
Synonyms:
1,2-Benzisothiazol-3-one
B2430-25G
B2430-500G
TCB2430-25GEA
114.51
USD
TCB2430-25G
TCB2430-500G
1,2-Benzisothiazol-3(2H)-one ≥98.0%
1,2-Benzisothiazol-3(2H)-one
CAS Number: 2634-33-5
MDL Number: MFCD00127753
Molecular Formula: C7H5NOS
Molecular Weight: 151.18
Purity/Analysis Method:
Formula:
C₇H₅NOS MW: 151.19 Melting Pt: 158 °C |
MDL Number:
MFCD00044001 CAS Number: 2634-33-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...